missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzophenone, 98%

Catalog No. p-7022128
Change view
Click to view available options
Quantity:
25 g
100 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1475514 25 g
AAA1475522 100 g
AAA1475530 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1475514 Supplier Thermo Scientific Chemicals Supplier No. A1475514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 90-90-4 | C13H9BrO | 261.12 g/mol

4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzophenone is used in analytical applications such as HPLC and GC. It is also used as a chemical reagent and pharmaceutical intermediate.

Solubility
Insoluble in water but slightly soluble in other solvents such as ethanol, ether and benzene.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from strong oxidizing agents.

Chemical Identifiers

CAS 90-90-4
Molecular Formula C13H9BrO
Molecular Weight (g/mol) 261.12
MDL Number MFCD00000103
InChI Key KEOLYBMGRQYQTN-UHFFFAOYSA-N
Synonym 4-bromobenzophenone, 4-bromophenyl phenyl methanone, p-bromobenzophenone, methanone, 4-bromophenyl phenyl, benzophenone, 4-bromo, p-benzoylbromobenzene, usaf do-3, 4-bromophenyl phenyl ketone, 4-bromophenyl-phenyl-methanone, 4-bromophenyl-phenylmethanone
PubChem CID 7030
SMILES BrC1=CC=C(C=C1)C(=O)C1=CC=CC=C1

Specifications

Melting Point 79°C to 83°C
Boiling Point 350°C
Odor Odorless
Quantity 25 g
Beilstein 1910182
Solubility Information Insoluble in water but slightly soluble in other solvents such as ethanol,ether and benzene.
IUPAC Name (4-bromophenyl)-phenylmethanone
Formula Weight 261.12
Percent Purity 98%
Chemical Name or Material 4-Bromobenzophenone
Hazard Category H315-H319-H335
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 202-024-9
RTECSNumber DJ0350000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only