missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromobenzo-18-crown 6-Ether 95.0+%, TCI America™

Catalog No. B21811G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21811G 1 g
B21815G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21811G Supplier TCI America Supplier No. B21811G

Chemical Identifiers

CAS 75460-28-5
Molecular Formula C16H23BrO6
Molecular Weight (g/mol) 391.258
MDL Number MFCD00143200
InChI Key XHHZIFBFFGIFNI-UHFFFAOYSA-N
Synonym 4'-bromobenzo-18-crown 6-ether, 4'-bromobenzo-18-crown-6, 4-bromobenzo-18-crown-6, 18-bromo-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine, 2,3-4-bromobenzo-1,4,7,10,13,16-hexaoxacyclooctadec-2-ene, 20-bromo-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene, 4-bromobenzo-3,4-18-crown-6, 2,3-4-bromobenzo-1,4,7,10,13,16-hexaoxacycloocta, 18-bromo-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecin, 20-bromanyl-2,5,8,11,14,17-hexaoxabicyclo 16.4.0 docosa-1 18 ,19,21-triene
PubChem CID 2734267
IUPAC Name 20-bromo-2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(18),19,21-triene
SMILES C1COCCOCCOC2=C(C=CC(=C2)Br)OCCOCCO1

Specifications

Melting Point 80°C
Color White-Yellow
Quantity 1 g
Formula Weight 391.26
Percent Purity ≥95.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromobenzo-18-crown 6-Ether