accessibility menu, dialog, popup

UserName

BrOOOOOO

CAS RN 75460-28-5

IUPAC Name: 18-bromo-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Synonyms: 18-bromo-2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Molecular Weight (g/mol): 391.26
Molecular Formula: C16H23BrO6
InChi Key: XHHZIFBFFGIFNI-UHFFFAOYSA-N
SMILES: BrC1=CC=C2OCCOCCOCCOCCOCCOC2=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results