missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzhydrol 97.0+%, TCI America™

Catalog No. B46805G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46805G 5 g
B46801G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46805G Supplier TCI America Supplier No. B46805G
Only null left

Chemical Identifiers

CAS 29334-16-5
Molecular Formula C13H11BrO
Molecular Weight (g/mol) 263.13
MDL Number MFCD00016851
InChI Key WTIWDBNPPSHSCB-UHFFFAOYNA-N
Synonym 4-Bromodiphenylmethanol
PubChem CID 98221
IUPAC Name (4-bromophenyl)(phenyl)methanol
SMILES OC(C1=CC=CC=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 65°C
Quantity 5 g
Formula Weight 263.13
Percent Purity ≥97.0% (GC)
Physical Form Powder
Chemical Name or Material 4-Bromobenzhydrol