missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromobenzenesulfonyl Chloride 98.0+%, TCI America™
Supplier: TCI America B0550500G
Specifications
| 4-Bromobenzenesulfonyl Chloride | |
| 75°C | |
| 153°C | |
| MFCD00007437 | |
| 3261 | |
| KMMHZIBWCXYAAH-UHFFFAOYSA-N | |
| 4-bromobenzenesulfonyl chloride | |
| 7396 | |
| ≥98.0% (GC,T) |
| 98-58-8 | |
| White-Yellow | |
| C6H4BrClO2S | |
| 500 g | |
| 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone | |
| C1=CC(=CC=C1S(=O)(=O)Cl)Br | |
| 255.51 | |
| 255.51 | |
| Crystalline Powder |
Chemical Identifiers
| 98-58-8 | |
| 255.51 | |
| KMMHZIBWCXYAAH-UHFFFAOYSA-N | |
| 7396 | |
| C1=CC(=CC=C1S(=O)(=O)Cl)Br |
| C6H4BrClO2S | |
| MFCD00007437 | |
| 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone | |
| 4-bromobenzenesulfonyl chloride |
Safety and Handling
TSCA : Yes