missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonyl Chloride 98.0+%, TCI America™

Catalog No. p-7118517
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B055025G 25 g
B0550500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B055025G Supplier TCI America Supplier No. B055025G
Only null left

Chemical Identifiers

CAS 98-58-8
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00007437
InChI Key KMMHZIBWCXYAAH-UHFFFAOYSA-N
Synonym 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone
PubChem CID 7396
IUPAC Name 4-bromobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)Br

Specifications

Melting Point 75°C
Color White-Yellow
Boiling Point 153°C
Quantity 25 g
UN Number 3261
Formula Weight 255.51
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromobenzenesulfonyl Chloride