Learn More
4-Bromobenzenesulfonyl chloride, 98+%
CAS: 98-58-8 | C6H4BrClO2S | 255.51 g/mol
Supplier: Thermo Scientific Chemicals A1590922
Description
4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.
Solubility
Soluble in Tetrahydrofuran, petroleum ether and 1,4-dioxane. Insoluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas.
Specifications
| 4-Bromobenzenesulfonyl chloride | |
| 73°C to 77°C | |
| C6H4BrClO2S | |
| 100 g | |
| 743518 | |
| 14,1407 | |
| Soluble in Tetrahydrofuran,petroleum ether and 1,4-dioxane. Insoluble in water. | |
| C1=CC(=CC=C1S(=O)(=O)Cl)Br | |
| 255.51 | |
| 255.52 |
| 98-58-8 | |
| 152°C to 154°C (26 mmHg) | |
| MFCD00007437 | |
| UN3261 | |
| Moisture sensitive | |
| 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone | |
| KMMHZIBWCXYAAH-UHFFFAOYSA-N | |
| 4-bromobenzenesulfonyl chloride | |
| 7396 | |
| ≥98% |
Chemical Identifiers
| 98-58-8 | |
| 255.51 | |
| KMMHZIBWCXYAAH-UHFFFAOYSA-N | |
| 7396 | |
| C1=CC(=CC=C1S(=O)(=O)Cl)Br |
| C6H4BrClO2S | |
| MFCD00007437 | |
| 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone | |
| 4-bromobenzenesulfonyl chloride |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H318
DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber : 202-683-2
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only