missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonyl chloride, 98+%

Catalog No. p-7022922
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1590914 25 g
AAA1590922 100 g
AAA1590936 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1590914 Supplier Thermo Scientific Chemicals Supplier No. A1590914
Only null left

CAS: 98-58-8 | C6H4BrClO2S | 255.51 g/mol

4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenesulfonyl chloride is used as activating agent in the synthesis of oligodeoxyribo- and oligoribo- nucleotides in solution. It also used in the synthesis of 4-(N-allylsulfamoyl)phenylboronic acid and in protection of amines as 4-bromobenzenesulfonamides.

Solubility
Soluble in Tetrahydrofuran, petroleum ether and 1,4-dioxane. Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Moisture sensitive. Store under inert gas.

Chemical Identifiers

CAS 98-58-8
Molecular Formula C6H4BrClO2S
Molecular Weight (g/mol) 255.51
MDL Number MFCD00007437
InChI Key KMMHZIBWCXYAAH-UHFFFAOYSA-N
Synonym 4-bromobenzene-1-sulfonyl chloride, p-bromobenzenesulfonyl chloride, 4-bromo-benzenesulfonyl chloride, p-bromophenylsulfonyl chloride, benzenesulfonyl chloride, 4-bromo, 4-bromobenzenesulphonyl chloride, 4-bromobenzenesulfonylchloride, unii-c6347t8fp1, 4-bromobenzene sulfonyl chloride, 4-bromophenyl chlorosulfone
PubChem CID 7396
IUPAC Name 4-bromobenzenesulfonyl chloride
SMILES C1=CC(=CC=C1S(=O)(=O)Cl)Br

Specifications

Melting Point 73°C to 77°C
Boiling Point 152°C to 154°C (26 mmHg)
Quantity 25 g
UN Number UN3261
Beilstein 743518
Sensitivity Moisture sensitive
Merck Index 14,1407
Solubility Information Soluble in Tetrahydrofuran,petroleum ether and 1,4-dioxane. Insoluble in water.
Formula Weight 255.52
Percent Purity ≥98%
Chemical Name or Material 4-Bromobenzenesulfonyl chloride
Show More Show Less
Hazard Category H314-H318
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-683-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only