missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenesulfonamide, 98%

Catalog No. p-7044882
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0860203 1 g
AAL0860206 5 g
AAL0860214 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAL0860203 Supplier Thermo Scientific Chemicals Supplier No. L0860203

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 701-34-8 | C6H6BrNO2S | 236.083 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 701-34-8
Molecular Formula C6H6BrNO2S
Molecular Weight (g/mol) 236.083
MDL Number MFCD00051977
InChI Key STYQHICBPYRHQK-UHFFFAOYSA-N
Synonym p-bromobenzenesulfonamide, benzenesulfonamide, p-bromo, benzenesulfonamide, 4-bromo, 4-bromobenzene-1-sulfonamide, 4-bromo-benzenesulfonamide, 4-bromobenzene sulfonamide, pbsa, 4-bromophenylsulfonamide, 4-bromo-phenylsulfonamide, wln: zswr de
PubChem CID 69696
IUPAC Name 4-bromobenzenesulfonamide
SMILES C1=CC(=CC=C1S(=O)(=O)N)Br

Specifications

Melting Point 164°C to 167°C
Quantity 1 g
Beilstein 2691657
Formula Weight 236.09
Percent Purity 98%
Chemical Name or Material 4-Bromobenzenesulfonamide
Hazard Category H302-H315-H319-H335
Hazard Statement GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
EINECSNumber 000-000-0
RTECSNumber DB0550000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only