missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzenediazonium tetrafluoroborate, 96%

Catalog No. p-7028383
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2567006 5 g
AAB2567014 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAB2567006 Supplier Thermo Scientific Chemicals Supplier No. B2567006

CAS: 673-40-5 | C6H4BBrF4N2 | 270.82 g/mol

4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromobenzenediazonium tetrafluoroborate was used in preparation of surface modified Au(111) electrode, 1-(4′-bromophenyl)-3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-heptadecafluorodec-1-ene.

Notes
Moisture sensitive. Store away from oxidizing agents, water/ moisture.

Chemical Identifiers

CAS 673-40-5
Molecular Formula C6H4BBrF4N2
Molecular Weight (g/mol) 270.82
MDL Number MFCD00011894
InChI Key FXTCQUCYDJZGGU-UHFFFAOYSA-N
Synonym 4-bromobenzenediazonium tetrafluoroborate, acmc-209nyn, 4-bromobenzene-1-diazonium; tetrafluoroboranuide, boron +3 cation; 4-bromobenzenediazonium; tetrafluoride
PubChem CID 2734810
IUPAC Name 4-bromobenzenediazonium;tetrafluoroborate
SMILES F[B-](F)(F)F.BrC1=CC=C(C=C1)[N+]#N

Specifications

Melting Point ∼135°C (decomposition)
Quantity 5 g
UN Number UN1759
Sensitivity Light and moisture sensitive
Formula Weight 270.83
Percent Purity 96%
Chemical Name or Material 4-Bromobenzenediazonium tetrafluoroborate
Hazard Category H302+H312+H332-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Hazard Class: 8; Packaging Group: II
EINECSNumber 211-608-2
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only