missing translation for 'onlineSavingsMsg'
Learn More

4-Bromobenzaldehyde diethyl acetal, 99%, Thermo Scientific™

Chemical Identifiers

CAS 34421-94-8
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.14
MDL Number MFCD01863514
InChI Key BFSNEBVTOODGHZ-UHFFFAOYSA-N
Synonym 4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
PubChem CID 688340
IUPAC Name 1-bromo-4-(diethoxymethyl)benzene
SMILES CCOC(OCC)C1=CC=C(Br)C=C1
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Packaging Price Quantity & Availability  
AC390020100
View Documents Promotion Details
N/A 10g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

34421-94-8
259.14
BFSNEBVTOODGHZ-UHFFFAOYSA-N
688340
CCOC(OCC)C1=CC=C(Br)C=C1
C11H15BrO2
MFCD01863514
4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
1-bromo-4-(diethoxymethyl)benzene

Specifications

34421-94-8
100.0
75°C (0.2mmHg)
Authentic
C11H15BrO2
BrC6H4CH(OCH2CH3)2
1.274
BFSNEBVTOODGHZ-UHFFFAOYSA-N
1-bromo-4-(diethoxymethyl)benzene
688340
99%
98.5
1.2740g/mL
105°C
98.5% min. (GC)
1.5120 to 1.5140 (20°C, 589nm)
MFCD01863514
4-bromobenzaldehyde diethyl acetal, 1-bromo-4-diethoxymethyl benzene, 4-bromobenzaldehydediethylacetal, benzene, 1-bromo-4-diethoxymethyl, 4-bromophenyl diethoxymethane, 4-bromobenzaldehyde diethylacetal, 4-bromo-alpha,alpha-diethoxytoluene, pubchem5372, acmc-1aigt, 4-bromobenzaldehydediethyl acetal
CCOC(OCC)C1=CC=C(Br)C=C1
259.14
259.14
4-Bromobenzaldehyde diethyl acetal
Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

SDS
Documents

Documents

RUO – Research Use Only