missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole 97.0+%, TCI America™

Catalog No. B054725G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B054725G 25 g
B0547100G 100 g
B0547500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B054725G Supplier TCI America Supplier No. B054725G

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00000097
InChI Key QJPJQTDYNZXKQF-UHFFFAOYSA-N
Synonym 4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
PubChem CID 7730
ChEBI CHEBI:47257
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Br

Specifications

Color Yellow
Boiling Point 215°C
Quantity 25 g
Formula Weight 187.04
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromoanisole
EINECSNumber (3)-4065
RTECSNumber BZ8501000
TSCA Yes