missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoanisole, 99%

Catalog No. p-7081729
Change view
Click to view available options
Quantity:
100 g
500 g
2500 g
Catalog No. Quantity
AAA1182422 100 g
AAA1182436 500 g
AAA118240E 2500 g
3 options

Catalog No. AAA1182422

Supplier: Thermo Scientific Chemicals A1182422

Only null left

CAS: 104-92-7 | C7H7BrO | 187.036 g/mol

4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromoanisole finds application as an intermediate in synthetic chemistry. It is used in the preparation of aryl 1,3-diketones and ethyl 4-methoxycinnamate. It is a used as a brominating reagent. Further, it is used in Suzuki coupling reaction with phenylboronic acid as well as in Heck reaction.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong acids.

Chemical Identifiers

CAS 104-92-7
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.036
MDL Number MFCD00000097
InChI Key QJPJQTDYNZXKQF-UHFFFAOYSA-N
Synonym 4-bromoanisole, p-bromoanisole, benzene, 1-bromo-4-methoxy, 4-methoxybromobenzene, anisyl bromide, p-anisyl bromide, p-bromanisole, p-methoxybromobenzene, p-methoxyphenyl bromide, 4-methoxyphenyl bromide
PubChem CID 7730
ChEBI CHEBI:47257
IUPAC Name 1-bromo-4-methoxybenzene
SMILES COC1=CC=C(C=C1)Br

Specifications

Melting Point 10°C to 13°C
Density 1.494
Boiling Point 98°C to 99°C (10 mm)
Flash Point 94°C (201°F)
Assay Percent Range 99%
Refractive Index 1.564
Quantity 100 g
Beilstein 1237590
Merck Index 14,1427
Solubility Information Immiscible with water.
Formula Weight 187.04
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 4-Bromoanisole
Show More Show Less
EINECSNumber 203-252-1
RTECSNumber BZ8501000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only