missing translation for 'onlineSavingsMsg'
Learn More

4-Bromoaniline 99.0+%, TCI America™

Supplier:  TCI America B1393500G

 View more versions of this product

Catalog No. B1393500G


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

4-Bromoaniline
65°C
124°C
MFCD00007822
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
C1=CC(=CC=C1N)Br
172.025
172.03
Crystalline Powder
106-40-1
White-Yellow
C6H6BrN
500 g
WDFQBORIUYODSI-UHFFFAOYSA-N
4-bromoaniline
7807
≥99.0% (GC)

Chemical Identifiers

106-40-1
172.025
WDFQBORIUYODSI-UHFFFAOYSA-N
7807
C1=CC(=CC=C1N)Br
C6H6BrN
MFCD00007822
p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine
4-bromoaniline
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

EINECSNumber : (3)-0221

RTECSNumber : BW9280000

TSCA : Yes

SDS
Documents

Documents

Promotions

Promotions