Learn More
4-Bromoaniline, 98+%
CAS: 106-40-1 | C6H6BrN | 172.025 g/mol
Supplier: Thermo Scientific Chemicals A151260B
Description
4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications4-Bromoaniline is used as a precursor in the production of azo dyes and dihydroquinazolines. It is also used as carbon and nitrogen supplement in the culture medium of Moraxella sp. strain G. Further, it is employed in the preparation of para-bromobiphenyl through Gomberg-Bachmann reaction. In addition to this, it is used as a building block in the preparation or pharmaceutical and organic compounds.
Solubility
Soluble in alcohol and ether. Insoluble in cold water.
Notes
Store in a cool place. Light sensitive and air sensitive. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with acids, acid chlorides, acid anhydrides, chloroformates and strong oxidizing agents.
Specifications
| 4-Bromoaniline | |
| 60°C to 65°C | |
| ≥98% | |
| C6H6BrN | |
| 1000 g | |
| 742031 | |
| p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine | |
| WDFQBORIUYODSI-UHFFFAOYSA-N | |
| 4-bromoaniline | |
| 7807 | |
| ≥98% |
| 106-40-1 | |
| 222°C to 224°C | |
| Characteristic | |
| MFCD00007822 | |
| UN2811 | |
| 14,1404 | |
| Soluble in alcohol and ether. Insoluble in cold water. | |
| C1=CC(=CC=C1N)Br | |
| 172.025 | |
| 172.03 |
Chemical Identifiers
| 106-40-1 | |
| 172.025 | |
| WDFQBORIUYODSI-UHFFFAOYSA-N | |
| 7807 | |
| C1=CC(=CC=C1N)Br |
| C6H6BrN | |
| MFCD00007822 | |
| p-bromoaniline, 4-bromobenzenamine, benzenamine, 4-bromo, p-bromophenylamine, aniline, p-bromo, 4-bromo-aniline, 4-bromobenzeneamine, p-bromo aniline, 4-bromanilinu, 4-bromo-phenylamine | |
| 4-bromoaniline |
Safety and Handling
GHS H Statement
H311-H373-H302-H332-H315-H319
Toxic in contact with skin.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
P260-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P314-P330-P332+P313-P361-P363-P501c
H302+H332-H311-H315-H319-H373
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.
EINECSNumber : 203-393-9
RTECSNumber : BW9280000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only