missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4'-Bromoacetanilide 98.0+%, TCI America™
Supplier: TCI America B0534100G
Specifications
| 4′-Bromoacetanilide | |
| 168°C | |
| MFCD00000092 | |
| 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide | |
| CC(=O)NC1=CC=C(Br)C=C1 | |
| 214.06 | |
| 214.06 | |
| Crystalline Powder |
| 103-88-8 | |
| C8H8BrNO | |
| 100 g | |
| MSLICLMCQYQNPK-UHFFFAOYSA-N | |
| N-(4-bromophenyl)acetamide | |
| 7683 | |
| ≥98.0% (GC) |
Chemical Identifiers
| 103-88-8 | |
| 214.06 | |
| MSLICLMCQYQNPK-UHFFFAOYSA-N | |
| 7683 | |
| CC(=O)NC1=CC=C(Br)C=C1 |
| C8H8BrNO | |
| MFCD00000092 | |
| 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide | |
| N-(4-bromophenyl)acetamide |
Safety and Handling
EINECSNumber : (3)-0196
RTECSNumber : AD9625000
TSCA : Yes