missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromoacetanilide 98.0+%, TCI America™

Catalog No. p-7118503
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B053425G 25 g
B0534100G 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B053425G Supplier TCI America Supplier No. B053425G
Only null left

Chemical Identifiers

CAS 103-88-8
Molecular Formula C8H8BrNO
Molecular Weight (g/mol) 214.06
MDL Number MFCD00000092
InChI Key MSLICLMCQYQNPK-UHFFFAOYSA-N
Synonym 4'-bromoacetanilide, n-4-bromophenyl acetamide, p-bromoacetanilide, 4-bromoacetanilide, bromoanilide, acetamide, n-4-bromophenyl, antisepsin, bromoantifebrin, asepsin, p-bromo-n-acetanilide
PubChem CID 7683
IUPAC Name N-(4-bromophenyl)acetamide
SMILES CC(=O)NC1=CC=C(Br)C=C1

Specifications

Melting Point 168°C
Quantity 25 g
Formula Weight 214.06
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromoacetanilide
EINECSNumber (3)-0196
RTECSNumber AD9625000
TSCA Yes