missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-p-terphenyl 97.0+%, TCI America™

Catalog No. B21551G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B21551G 1 g
B21555G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B21551G Supplier TCI America Supplier No. B21551G

Chemical Identifiers

CAS 1762-84-1
Molecular Formula C18H13Br
Molecular Weight (g/mol) 309.206
MDL Number MFCD02685117
InChI Key XNBGHWRYLJOQAU-UHFFFAOYSA-N
PubChem CID 11012297
IUPAC Name 1-bromo-4-(4-phenylphenyl)benzene
SMILES C1=CC=C(C=C1)C2=CC=C(C=C2)C3=CC=C(C=C3)Br

Specifications

Melting Point 232°C
Color White
Quantity 1 g
UN Number 3152
Formula Weight 309.21
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-p-terphenyl