missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-o-xylene (contains 3-Bromo-o-xylene) 75.0+%, TCI America™

Catalog No. B134625G
Change view
Click to view available options
Quantity:
25 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B134625G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B134625G Supplier TCI America Supplier No. B134625G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Laboratory reagent grade 4-bromo-o-xylene, typically used as a chemical building block.

Specifications

Melting Point 0°C
CAS 583-71-1
Color Yellow
Quantity 25 g
Physical Form Clear Liquid at 20°C
Molecular Formula C8H9Br
MDL Number MFCD00000086
UN Number 3082
Synonym 4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
Boiling Point 208°C
InChI Key QOGHRLGTXVMRLM-UHFFFAOYSA-N
SMILES CC1=C(C=C(C=C1)Br)C
IUPAC Name 4-bromo-1,2-dimethylbenzene
Molecular Weight (g/mol) 185.064
PubChem CID 68504
Formula Weight 185.06
Percent Purity ≥75.0% (GC)
Grade Not Available
Chemical Name or Material 4-Bromo-o-xylene (contains 3-Bromo-o-xylene)
Show More Show Less
RTECSNumber CY9020400
TSCA Yes