missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-o-xylene, 97%

Catalog No. AAA1473622
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1473622 100 g
AAA1473606 5 g
AAA1473614 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1473622 Supplier Thermo Scientific Chemicals Supplier No. A1473622
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 583-71-1 | C8H9Br | 185.064 g/mol

4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Bromo-o-xylene is used as synthetic organic materials and soluble polyimide resin intermediates.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Chemical Identifiers

CAS 583-71-1
Molecular Formula C8H9Br
Molecular Weight (g/mol) 185.064
MDL Number MFCD00000086
InChI Key QOGHRLGTXVMRLM-UHFFFAOYSA-N
Synonym 4-bromo-o-xylene, 3,4-dimethylbromobenzene, 1-bromo-3,4-dimethylbenzene, benzene, 4-bromo-1,2-dimethyl, 3,4-xylyl bromide, 4-bromo-1,2-xylene, 4-bromo-1,2-dimethyl-benzene, 3,4-dimethylphenyl bromide, 4-bromo-ortho-xylene, o-xylene, 4-bromo
PubChem CID 68504
IUPAC Name 4-bromo-1,2-dimethylbenzene
SMILES CC1=C(C=C(C=C1)Br)C

Specifications

Density 1.37
Quantity 100 g
Refractive Index 1.556
Beilstein 2040354
Boiling Point 214°C to 215°C
Solubility Information Not miscible in water.
Formula Weight 185.07
Percent Purity 97%
Chemical Name or Material 4-Bromo-o-xylene
Hazard Category H227-H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Precautionary Statement P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
EINECSNumber 209-518-3
RTECSNumber CY9020400
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only