missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-o-cresol 98.0+%, TCI America™

Catalog No. p-7123097
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36775G 5 g
B367725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36775G Supplier TCI America Supplier No. B36775G
Only null left

Chemical Identifiers

CAS 2362-12-1
Molecular Formula C7H7BrO
Molecular Weight (g/mol) 187.04
MDL Number MFCD00055435
InChI Key IWJGMJHAIUBWKT-UHFFFAOYSA-N
Synonym 4-bromo-o-cresol, 2-methyl-4-bromophenol, 4-bromo-2-methyl-phenol, o-cresol, 4-bromo, phenol, 4-bromo-2-methyl, 2-hydroxy-5-bromotoluene, p-bromocresol, 4-bromo-cresol, pubchem7106, acmc-1chy8
PubChem CID 16896
IUPAC Name 4-bromo-2-methylphenol
SMILES CC1=C(C=CC(=C1)Br)O

Specifications

Melting Point 66°C
Boiling Point 88°C
Quantity 5 g
Formula Weight 187.04
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-o-cresol
RTECSNumber GO6868000
TSCA No