missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N-dimethylaniline 98.0+%, TCI America™

Catalog No. p-7118542
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
Catalog No. Quantity
B058525G 25 g
B0585100G 100 g
B0585500G 500 g
3 options

Catalog No. B058525G

Supplier: TCI America B058525G

Only null left

Chemical Identifiers

CAS 586-77-6
Molecular Formula C8H10BrN
Molecular Weight (g/mol) 200.08
MDL Number MFCD00000093
InChI Key XYZWMVYYUIMRIZ-UHFFFAOYSA-N
Synonym n,n-dimethyl-p-bromoaniline, 4-dimethylaminobromobenzene, p-bromo-n,n-dimethylaniline, benzenamine, 4-bromo-n,n-dimethyl, n,n-dimethyl-4-bromoaniline, p-dimethylamino phenyl bromide, p-dimethylaminobromobenzene, 1-bromo-4-dimethylamino benzene, p-bromo dimethylamino benzene, p-n,n-dimethylaminobromobenzene
PubChem CID 11465
IUPAC Name 4-bromo-N,N-dimethylaniline
SMILES CN(C)C1=CC=C(Br)C=C1

Specifications

Melting Point 54°C
Color White
Boiling Point 264°C
Quantity 25 g
Formula Weight 200.08
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-N,N-dimethylaniline
RTECSNumber BW9300000
TSCA Yes