missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N-diethylaniline 97.0+%, TCI America™

Catalog No. B30275G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B30275G 5 g
B302725G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B30275G Supplier TCI America Supplier No. B30275G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 2052-06-4
Molecular Formula C10H14BrN
Molecular Weight (g/mol) 228.13
MDL Number MFCD00013530
InChI Key NGYMZFJVHHKJQR-UHFFFAOYSA-N
Synonym p-bromo-n,n-diethylaniline, benzenamine, 4-bromo-n,n-diethyl, n,n-diethyl-p-bromoaniline, 4-n,n-diethylamino-bromobenzene, benzenamine,4-bromo-n,n-diethyl, p-n,nddiethylaniline, 4-bromodiethylaniline, 4-bromophenyldiethylamine, 4-bromophenyl diethylamine, 4-bromo-phenyl diethylamine
PubChem CID 16328
IUPAC Name 4-bromo-N,N-diethylaniline
SMILES CCN(CC)C1=CC=C(C=C1)Br

Specifications

Color White-Yellow
Boiling Point 215°C
Quantity 5 g
Formula Weight 228.13
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 4-Bromo-N,N-diethylaniline
TSCA Yes
Recommended Storage Refrigerator