accessibility menu, dialog, popup

UserName

CH3NCH3Br

CAS RN 2052-06-4

IUPAC Name: 4-bromo-N,N-diethylaniline
Synonyms: 4-bromo-N,N-diethylaniline
Molecular Weight (g/mol): 228.13
Molecular Formula: C10H14BrN
InChi Key: NGYMZFJVHHKJQR-UHFFFAOYSA-N
SMILES: CCN(CC)C1=CC=C(Br)C=C1
Boiling Point 
  • (2)
  • (1)
Percent Purity 
  • (2)
  • (1)
Quantity 
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)