missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N,N,3-trimethylaniline 98.0+%, TCI America™

Catalog No. B31335G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B31335G 5 g
B313325G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B31335G Supplier TCI America Supplier No. B31335G

Chemical Identifiers

CAS 50638-50-1
Molecular Formula C9H12BrN
Molecular Weight (g/mol) 214.106
MDL Number MFCD01851099
InChI Key DPFVFQHTPIFECX-UHFFFAOYSA-N
Synonym 4-Bromo-N,N-dimethyl-m-toluidine
PubChem CID 12385715
IUPAC Name 4-bromo-N,N,3-trimethylaniline
SMILES CC1=C(C=CC(=C1)N(C)C)Br

Specifications

Melting Point 54°C
Color White-Yellow
Boiling Point 148°C
Quantity 5 g
Formula Weight 214.11
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-N,N,3-trimethylaniline