missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylglycine, 95%, Thermo Scientific Chemicals

Catalog No. p-7032418
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH3386503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AAH3386503 Supplier Thermo Scientific Chemicals Supplier No. H3386503

CAS: 119397-06-7 | Molecular Formula: C8H8BrNO2 | Molecular Weight: 230.06 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 119397-06-7
Molecular Formula C8H8BrNO2
Molecular Weight (g/mol) 230.06
MDL Number MFCD00049327
InChI Key APLQICUORRMFHY-UHFFFAOYNA-N
Synonym 2-amino-2-4-bromophenyl acetic acid, amino 4-bromophenyl acetic acid, amino-4-bromo-phenyl-acetic acid, 2-4-bromophenyl glycine, 4-bromo-dl-phenylglycine, 2-amino-2-4-bromophenyl aceticacid, dl-4-br-phg-oh, 2-4-bromophenyl-dl-glycine, +-2-amino-2-4-bromophenyl acetic acid
PubChem CID 2764157
IUPAC Name 2-amino-2-(4-bromophenyl)acetic acid
SMILES NC(C(O)=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 273°C to 274°C
Quantity 1 g
Formula Weight 230.1
Percent Purity 95%
Chemical Name or Material 4-Bromo-DL-phenylglycine
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only