accessibility menu, dialog, popup

UserName

CAS RN 119397-06-7

IUPAC Name: 2-amino-2-(4-bromophenyl)acetic acid
Synonyms: amino(4-bromophenyl)acetic acid
Molecular Weight (g/mol): 230.06
Molecular Formula: C8H8BrNO2
InChi Key: APLQICUORRMFHY-UHFFFAOYNA-N
SMILES: NC(C(O)=O)C1=CC=C(Br)C=C1

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (1)
  • (1)
  • (1)