missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-DL-phenylalanine, 98+%, Thermo Scientific Chemicals

Catalog No. p-7043807
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0283303 1 g
AAL0283306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0283303 Supplier Thermo Scientific Chemicals Supplier No. L0283303

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14091-15-7
Molecular Formula C9H10BrNO2
Molecular Weight (g/mol) 244.09
MDL Number MFCD00002599
InChI Key PEMUHKUIQHFMTH-UHFFFAOYSA-N
Synonym 2-amino-3-4-bromophenyl propanoic acid, p-bromo-dl-phenylalanine, 4-bromo-dl-phenylalanine, dl-4-br-phe-oh, 4-bromophenylalanine, 4-bromo-phenylalanine, h-dl-phe 4-br-oh, 2-amino-3-4-bromophenyl propionic acid, 2-amino-3-4-bromo-phenyl-propionic acid, p-br-phenylalanine
PubChem CID 85681
IUPAC Name 2-amino-3-(4-bromophenyl)propanoic acid
SMILES NC(CC1=CC=C(Br)C=C1)C(O)=O

Specifications

Melting Point ∽262°C (decomposition)
Beilstein 2212158
Quantity 1 g
Odor Odorless
Formula Weight 244.09
Percent Purity ≥98%
Chemical Name or Material 4-Bromo-DL-phenylalanine
EINECSNumber 237-941-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only