missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-alpha-methylbenzyl Alcohol 99.0+%, TCI America™
$94.33 - $265.67
Chemical Identifiers
| CAS | 5391-88-8 |
|---|---|
| Molecular Formula | C8H9BrO |
| Molecular Weight (g/mol) | 201.06 |
| MDL Number | MFCD00004514,MFCD06201860,MFCD06201861 |
| InChI Key | XTDTYSBVMBQIBT-UHFFFAOYNA-N |
| Synonym | 1-4-bromophenyl ethanol, 4-bromo-alpha-methylbenzyl alcohol, 1-4-bromophenyl ethan-1-ol, 4-bromophenyl methyl carbinol, p-bromophenylmethylcarbinol, 4-bromophenylmethylcarbinol, 4-bromo-a-methylbenzyl alcohol, benzenemethanol, 4-bromo-alpha-methyl, benzyl alcohol, p-bromo-.alpha.-methyl, benzenemethanol, 4-bromo-.alpha.-methyl |
| PubChem CID | 95352 |
| IUPAC Name | 1-(4-bromophenyl)ethan-1-ol |
| SMILES | CC(O)C1=CC=C(Br)C=C1 |
Chemical Identifiers
| 5391-88-8 | |
| 201.06 | |
| XTDTYSBVMBQIBT-UHFFFAOYNA-N | |
| 95352 | |
| CC(O)C1=CC=C(Br)C=C1 |
| C8H9BrO | |
| MFCD00004514,MFCD06201860,MFCD06201861 | |
| 1-4-bromophenyl ethanol, 4-bromo-alpha-methylbenzyl alcohol, 1-4-bromophenyl ethan-1-ol, 4-bromophenyl methyl carbinol, p-bromophenylmethylcarbinol, 4-bromophenylmethylcarbinol, 4-bromo-a-methylbenzyl alcohol, benzenemethanol, 4-bromo-alpha-methyl, benzyl alcohol, p-bromo-.alpha.-methyl, benzenemethanol, 4-bromo-.alpha.-methyl | |
| 1-(4-bromophenyl)ethan-1-ol |
Specifications
| 5391-88-8 | |
| 134°C | |
| MFCD00004514,MFCD06201860,MFCD06201861 | |
| 1-4-bromophenyl ethanol, 4-bromo-alpha-methylbenzyl alcohol, 1-4-bromophenyl ethan-1-ol, 4-bromophenyl methyl carbinol, p-bromophenylmethylcarbinol, 4-bromophenylmethylcarbinol, 4-bromo-a-methylbenzyl alcohol, benzenemethanol, 4-bromo-alpha-methyl, benzyl alcohol, p-bromo-.alpha.-methyl, benzenemethanol, 4-bromo-.alpha.-methyl | |
| CC(O)C1=CC=C(Br)C=C1 | |
| 201.06 | |
| 201.06 | |
| Crystalline Lumps |
| White | |
| C8H9BrO | |
| 5 g | |
| XTDTYSBVMBQIBT-UHFFFAOYNA-N | |
| 1-(4-bromophenyl)ethan-1-ol | |
| 95352 | |
| ≥99.0% (GC) | |
| 4-Bromo-alpha-methylbenzyl Alcohol |
Safety and Handling
TSCA : No