missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-alpha-methylbenzyl Alcohol 99.0+%, TCI America™
Specifications
Specifications
| Color | White |
| Boiling Point | 134°C |
| Quantity | 5 g |
| Formula Weight | 201.06 |
| Percent Purity | ≥99.0% (GC) |
| Physical Form | Crystalline Lumps |
| Chemical Name or Material | 4-Bromo-alpha-methylbenzyl Alcohol |
Safety and Handling
| TSCA | No |