missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™
$67.11 - $221.83
Chemical Identifiers
| CAS | 1161009-88-6 |
|---|---|
| Molecular Formula | C25H15Br |
| Molecular Weight (g/mol) | 395.299 |
| MDL Number | MFCD16660920 |
| InChI Key | GQXFSXMUBDPXBG-UHFFFAOYSA-N |
| PubChem CID | 68018887 |
| IUPAC Name | 4-bromo-9,9'-spirobi[fluorene] |
| SMILES | C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=C(C6=CC=CC=C46)C(=CC=C5)Br |
Chemical Identifiers
| 1161009-88-6 | |
| 395.299 | |
| GQXFSXMUBDPXBG-UHFFFAOYSA-N | |
| 4-bromo-9,9'-spirobi[fluorene] |
| C25H15Br | |
| MFCD16660920 | |
| 68018887 | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=C(C6=CC=CC=C46)C(=CC=C5)Br |
Specifications
| 1161009-88-6 | |
| White | |
| MFCD16660920 | |
| GQXFSXMUBDPXBG-UHFFFAOYSA-N | |
| 4-bromo-9,9'-spirobi[fluorene] | |
| 68018887 | |
| ≥98.0% (GC) | |
| 4-Bromo-9,9′-spirobi[9H-fluorene] |
| 159°C | |
| C25H15Br | |
| 1 g | |
| C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=C(C6=CC=CC=C46)C(=CC=C5)Br | |
| 395.299 | |
| 395.30 | |
| Crystalline Powder |
Safety and Handling
TSCA : No