accessibility menu, dialog, popup

UserName

Br

CAS RN 1161009-88-6

IUPAC Name: 5-bromo-9,9'-spirobi[fluorene]
Synonyms: 5-bromo-9,9'-spirobi[fluorene]
Molecular Weight (g/mol): 395.3
Molecular Formula: C25H15Br
InChi Key: GQXFSXMUBDPXBG-UHFFFAOYSA-N
SMILES: BrC1=CC=CC2=C1C1=CC=CC=C1C21C2=CC=CC=C2C2=CC=CC=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results