missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-9,9'-spirobi[9H-fluorene] 98.0+%, TCI America™

Catalog No. B50315G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B50315G 5 g
B50311G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B50315G Supplier TCI America Supplier No. B50315G

Chemical Identifiers

CAS 1161009-88-6
Molecular Formula C25H15Br
Molecular Weight (g/mol) 395.299
MDL Number MFCD16660920
InChI Key GQXFSXMUBDPXBG-UHFFFAOYSA-N
PubChem CID 68018887
IUPAC Name 4-bromo-9,9'-spirobi[fluorene]
SMILES C1=CC=C2C(=C1)C3=CC=CC=C3C24C5=C(C6=CC=CC=C46)C(=CC=C5)Br

Specifications

Melting Point 159°C
Color White
Quantity 5 g
Formula Weight 395.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-9,9′-spirobi[9H-fluorene]