missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-9,9-dimethylfluorene 98.0+%, TCI America™
$39.14 - $124.89
Chemical Identifiers
| CAS | 942615-32-9 |
|---|---|
| Molecular Formula | C15H13Br |
| Molecular Weight (g/mol) | 273.173 |
| MDL Number | MFCD23099415 |
| InChI Key | SXOUNESKHJIXNK-UHFFFAOYSA-N |
| Synonym | 4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene |
| PubChem CID | 59182836 |
| IUPAC Name | 4-bromo-9,9-dimethylfluorene |
| SMILES | CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B4661200MG
|
TCI America
B4661200MG |
200 mg |
Each for $39.14
|
|
|||||
|
B46611G
|
TCI America
B46611G |
1 g |
Each for $124.89
|
|
|||||
Chemical Identifiers
| 942615-32-9 | |
| 273.173 | |
| SXOUNESKHJIXNK-UHFFFAOYSA-N | |
| 59182836 | |
| CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C |
| C15H13Br | |
| MFCD23099415 | |
| 4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene | |
| 4-bromo-9,9-dimethylfluorene |
Specifications
| 942615-32-9 | |
| C15H13Br | |
| 200 mg | |
| SXOUNESKHJIXNK-UHFFFAOYSA-N | |
| 4-bromo-9,9-dimethylfluorene | |
| 59182836 | |
| ≥98.0% (GC) | |
| 4-Bromo-9,9-dimethylfluorene |
| White-Yellow | |
| MFCD23099415 | |
| 4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene | |
| CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C | |
| 273.173 | |
| 273.17 | |
| Crystalline Powder |
Safety and Handling
TSCA : No