missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-9,9-dimethylfluorene 98.0+%, TCI America™

Catalog No. B4661200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B4661200MG 200 mg
B46611G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B4661200MG Supplier TCI America Supplier No. B4661200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 942615-32-9
Molecular Formula C15H13Br
Molecular Weight (g/mol) 273.173
MDL Number MFCD23099415
InChI Key SXOUNESKHJIXNK-UHFFFAOYSA-N
Synonym 4-bromo-9,9-dimethyl-9h-fluorene, 4-bromo-9,9-dimethyl fluorene, 4-bromo-9,9-dimethyl-fluorene
PubChem CID 59182836
IUPAC Name 4-bromo-9,9-dimethylfluorene
SMILES CC1(C2=C(C3=CC=CC=C31)C(=CC=C2)Br)C

Specifications

Color White-Yellow
Quantity 200 mg
Formula Weight 273.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-9,9-dimethylfluorene