missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-5'-phenyl-1,1':3',1″-terphenyl 98.0+%, TCI America™

Catalog No. p-7123819
Change view
Click to view available options
Quantity:
1 g
5 g
Catalog No. Quantity
B48891G 1 g
B48895G 5 g
2 options

Catalog No. B48891G

Supplier: TCI America B48891G

Only null left

Chemical Identifiers

CAS 116941-52-7
Molecular Formula C24H17Br
Molecular Weight (g/mol) 385.304
MDL Number MFCD26407506
InChI Key SIAJAYFLPBYCOF-UHFFFAOYSA-N
PubChem CID 14022731
IUPAC Name 1-(4-bromophenyl)-3,5-diphenylbenzene
SMILES C1=CC=C(C=C1)C2=CC(=CC(=C2)C3=CC=C(C=C3)Br)C4=CC=CC=C4

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 1 g
Synonym 5′C-(4-Bromophenyl)-1,1′C:3′C,1′C′C-terphenyl, 4-Bromo-5′C-phenyl-m-terphenyl, 5′C-(4-Bromophenyl)-m-terphenyl
Formula Weight 385.30
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-5′-phenyl-1,1′:3′,1′′-terphenyl