missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-methylbiphenyl 98.0+%, TCI America™

Catalog No. B48041G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B48041G 1 g
B48045G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B48041G Supplier TCI America Supplier No. B48041G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 50670-49-0
Molecular Formula C13H11Br
Molecular Weight (g/mol) 247.14
MDL Number MFCD00466236
InChI Key MYWLXURJCXISCT-UHFFFAOYSA-N
Synonym 4-bromo-4'-methylbiphenyl, 4-bromo-4'-methyl-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-methyl, biphenyl, 4-bromo-4'-methyl, 4-bromo-4'-methyl-biphenyl, 1-bromo-4-4-methylphenyl benzene, 1-4-bromophenyl-4-methylbenzene, 4-brromo-4-methylbiphenyl, 4-brom-4'-methyl-biphenyl, 4'-bromo-4-methyl-biphenyl
PubChem CID 521291
IUPAC Name 4-bromo-4'-methyl-1,1'-biphenyl
SMILES CC1=CC=C(C=C1)C1=CC=C(Br)C=C1

Specifications

Melting Point 133°C
Color White-Yellow
Boiling Point 315°C
Quantity 1 g
Formula Weight 247.14
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-methylbiphenyl