missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-iodobiphenyl 98.0+%, TCI America™

Catalog No. p-7123073
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36485G 5 g
B364825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36485G Supplier TCI America Supplier No. B36485G

Chemical Identifiers

CAS 105946-82-5
Molecular Formula C12H8BrI
Molecular Weight (g/mol) 359.004
InChI Key GWOAJJWBCSUGHH-UHFFFAOYSA-N
Synonym 4-bromo-4'-iodobiphenyl, 4-bromo-4'-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-iodo, 1-bromo-4-4-iodophenyl benzene, 4'-bromo-4-iodobiphenyl, 4-iodo-4'-bromobiphenyl, 4-bromo-4'-iodo-biphenyl, 4-bromo-4/'-iodobiphenyl, ksc164g2f
PubChem CID 10904452
IUPAC Name 1-bromo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)Br

Specifications

Melting Point 177°C
Color White-Yellow
Quantity 5 g
UN Number 3152
Formula Weight 359.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-4′-iodobiphenyl