missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-iodobiphenyl, 98%

Catalog No. p-7055707
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6435514 25 g
AAH6435522 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6435514 Supplier Thermo Scientific Chemicals Supplier No. H6435514

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 105946-82-5 | C12H8BrI | 359.004 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 105946-82-5
Molecular Formula C12H8BrI
Molecular Weight (g/mol) 359.004
MDL Number MFCD17676204
InChI Key GWOAJJWBCSUGHH-UHFFFAOYSA-N
Synonym 4-bromo-4'-iodobiphenyl, 4-bromo-4'-iodo-1,1'-biphenyl, 1,1'-biphenyl, 4-bromo-4'-iodo, 1-bromo-4-4-iodophenyl benzene, 4'-bromo-4-iodobiphenyl, 4-iodo-4'-bromobiphenyl, 4-bromo-4'-iodo-biphenyl, 4-bromo-4/'-iodobiphenyl, ksc164g2f
PubChem CID 10904452
IUPAC Name 1-bromo-4-(4-iodophenyl)benzene
SMILES C1=CC(=CC=C1C2=CC=C(C=C2)I)Br

Specifications

Melting Point 175°C to 179°C (literature)
Quantity 25 g
UN Number UN3152
Beilstein 2518914
Sensitivity Light sensitive
Formula Weight 359
Percent Purity 98%
Physical Form Solid
Chemical Name or Material 4-Bromo-4'-iodobiphenyl
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
DOTInformation Transport Hazard Class: 9; Packaging Group: II; Proper Shipping Name: POLYHALOGENATED BIPHENYLS, SOLID
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only