missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-4'-chlorobenzophenone 98.0+%, TCI America™
$277.75 - $963.71
Chemical Identifiers
| CAS | 27428-57-5 |
|---|---|
| Molecular Formula | C13H8BrClO |
| Molecular Weight (g/mol) | 295.56 |
| MDL Number | MFCD00045208 |
| InChI Key | FYMCXGILCMIOKD-UHFFFAOYSA-N |
| PubChem CID | 2756868 |
| IUPAC Name | (4-bromophenyl)(4-chlorophenyl)methanone |
| SMILES | ClC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1 |
Chemical Identifiers
| 27428-57-5 | |
| 295.56 | |
| FYMCXGILCMIOKD-UHFFFAOYSA-N | |
| (4-bromophenyl)(4-chlorophenyl)methanone |
| C13H8BrClO | |
| MFCD00045208 | |
| 2756868 | |
| ClC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1 |
Specifications
| 27428-57-5 | |
| White-Yellow | |
| MFCD00045208 | |
| FYMCXGILCMIOKD-UHFFFAOYSA-N | |
| (4-bromophenyl)(4-chlorophenyl)methanone | |
| 2756868 | |
| ≥98.0% (GC) | |
| 4-Bromo-4′-chlorobenzophenone |
| 150°C | |
| C13H8BrClO | |
| 1 g | |
| ClC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1 | |
| 295.56 | |
| 295.56 | |
| Powder |
Safety and Handling
TSCA : No