missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-4'-chlorobenzophenone 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 150°C |
| Color | White-Yellow |
| Quantity | 5 g |
| Formula Weight | 295.56 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Powder |
| Chemical Name or Material | 4-Bromo-4′-chlorobenzophenone |
Safety and Handling
| TSCA | No |