missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-4'-chlorobenzophenone 98.0+%, TCI America™

Catalog No. B42241G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B42241G 1 g
B42245G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B42241G Supplier TCI America Supplier No. B42241G
Only null left

Chemical Identifiers

CAS 27428-57-5
Molecular Formula C13H8BrClO
Molecular Weight (g/mol) 295.56
MDL Number MFCD00045208
InChI Key FYMCXGILCMIOKD-UHFFFAOYSA-N
PubChem CID 2756868
IUPAC Name (4-bromophenyl)(4-chlorophenyl)methanone
SMILES ClC1=CC=C(C=C1)C(=O)C1=CC=C(Br)C=C1

Specifications

Melting Point 150°C
Color White-Yellow
Quantity 1 g
Formula Weight 295.56
Percent Purity ≥98.0% (GC)
Physical Form Powder
Chemical Name or Material 4-Bromo-4′-chlorobenzophenone