missing translation for 'onlineSavingsMsg'
Learn More

4'-Bromo-4-biphenylboronic Acid (contains varying amounts of Anhydride), TCI America™

Catalog No. B28601G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B28601G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. B28601G Supplier TCI America Supplier No. B28601G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 480996-05-2
Molecular Formula C12H10BBrO2
Molecular Weight (g/mol) 276.924
MDL Number MFCD02251652
InChI Key HQAGDFJHKPSZHT-UHFFFAOYSA-N
Synonym 4-(4-Bromophenyl)phenylboronic Acid, 4-(4-Bromophenyl)benzeneboronic Acid
PubChem CID 3841607
IUPAC Name [4-(4-bromophenyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C2=CC=C(C=C2)Br)(O)O

Specifications

Color White-Yellow
Quantity 1 g
Formula Weight 276.92
Physical Form Crystalline Powder
Chemical Name or Material 4′-Bromo-4-biphenylboronic Acid (contains varying amounts of Anhydride)