missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrotoluene 96.0+%, TCI America™

Catalog No. p-7119307
Change view
Click to view available options
Quantity:
5 g
25 g
Catalog No. Quantity
B18545G 5 g
B185425G 25 g
2 options

Catalog No. B18545G

Supplier: TCI America B18545G

Only null left

Chemical Identifiers

CAS 5326-34-1
Molecular Formula C7H6BrNO2
Molecular Weight (g/mol) 216.03
MDL Number MFCD00024180
InChI Key UPBUTKQMDPHQAQ-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrotoluene, 2-bromo-5-methylnitrobenzene, benzene, 1-bromo-4-methyl-2-nitro, bromonitrotoluene-4, 1-bromo-4-methyl-2-nitro-benzene, 3-nitro-4-bromotoluene, pubchem9585, acmc-209l5d, ksc490m7t, tpc-i121
PubChem CID 79224
IUPAC Name 1-bromo-4-methyl-2-nitrobenzene
SMILES CC1=CC=C(Br)C(=C1)[N+]([O-])=O

Specifications

Boiling Point 152°C
Quantity 5 g
Formula Weight 216.03
Percent Purity ≥96.0% (GC)
Physical Form Crystals
Chemical Name or Material 4-Bromo-3-nitrotoluene