missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrobenzotrifluoride 98.0+%, TCI America™

Catalog No. B307425G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B307425G 25 g
B30745G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B307425G Supplier TCI America Supplier No. B307425G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 349-03-1
Molecular Formula C7H3BrF3NO2
Molecular Weight (g/mol) 270.01
MDL Number MFCD00014685
InChI Key PESPBNYBZVIGRO-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrobenzotrifluoride, 1-bromo-2-nitro-4-trifluoromethyl benzene, 2-nitro-4-trifluoromethylbromobenzene, 3-nitro-4-bromobenzotrifluoride, 2-bromo-5-trifluoromethyl nitrobenzene, 4-bromo-3-nitro-a,a,a-trifluorotoluene, benzene, 1-bromo-2-nitro-4-trifluoromethyl, 1-bromo-2-nitro-4-trifluoromethyl-benzene, 2-bromo-1-nitro-5-trifluoromethyl benzene, toluene, 4-bromo-3-nitro-alpha,alpha,alpha-trifluoro
PubChem CID 520706
IUPAC Name 1-bromo-2-nitro-4-(trifluoromethyl)benzene
SMILES [O-][N+](=O)C1=CC(=CC=C1Br)C(F)(F)F

Specifications

Color Yellow
Boiling Point 105°C
Quantity 25 g
Formula Weight 270.01
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 4-Bromo-3-nitrobenzotrifluoride