missing translation for 'onlineSavingsMsg'
Learn More
Learn More
4-Bromo-3-nitrobenzoic Acid 98.0+%, TCI America™
$124.89 - $493.97
Chemical Identifiers
| CAS | 6319-40-0 |
|---|---|
| Molecular Formula | C7H4BrNO4 |
| Molecular Weight (g/mol) | 246.016 |
| MDL Number | MFCD00272137 |
| InChI Key | RVCTZJVBWNFYRU-UHFFFAOYSA-N |
| Synonym | 4-bromo-3-nitrobenzoic acid, 3-nitro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-nitro, 4-bromo-3-nitro-benzoic acid, zlchem 531, pubchem2194, acmc-209ndq, ksc353s1b, 4-bromo-3-nitrobenzoic acid # |
| PubChem CID | 232986 |
| SMILES | C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Br |
Chemical Identifiers
| 6319-40-0 | |
| 246.016 | |
| RVCTZJVBWNFYRU-UHFFFAOYSA-N | |
| 232986 |
| C7H4BrNO4 | |
| MFCD00272137 | |
| 4-bromo-3-nitrobenzoic acid, 3-nitro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-nitro, 4-bromo-3-nitro-benzoic acid, zlchem 531, pubchem2194, acmc-209ndq, ksc353s1b, 4-bromo-3-nitrobenzoic acid # | |
| C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Br |
Specifications
| 6319-40-0 | |
| White-Yellow | |
| MFCD00272137 | |
| 4-bromo-3-nitrobenzoic acid, 3-nitro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-nitro, 4-bromo-3-nitro-benzoic acid, zlchem 531, pubchem2194, acmc-209ndq, ksc353s1b, 4-bromo-3-nitrobenzoic acid # | |
| C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Br | |
| 232986 | |
| ≥98.0% (GC,T) | |
| 4-Bromo-3-nitrobenzoic Acid |
| 204°C | |
| C7H4BrNO4 | |
| 5 g | |
| RVCTZJVBWNFYRU-UHFFFAOYSA-N | |
| 246.016 | |
| 246.02 | |
| Crystalline Powder |
Safety and Handling
TSCA : No