missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrobenzoic Acid 98.0+%, TCI America™

Catalog No. B36895G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B36895G 5 g
B368925G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B36895G Supplier TCI America Supplier No. B36895G

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6319-40-0
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00272137
InChI Key RVCTZJVBWNFYRU-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrobenzoic acid, 3-nitro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-nitro, 4-bromo-3-nitro-benzoic acid, zlchem 531, pubchem2194, acmc-209ndq, ksc353s1b, 4-bromo-3-nitrobenzoic acid #
PubChem CID 232986
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Br

Specifications

Melting Point 204°C
Color White-Yellow
Quantity 5 g
Formula Weight 246.02
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-nitrobenzoic Acid