missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-nitrobenzoic acid, 95%

Catalog No. AAH6197506
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH6197506 5 g
AAH6197514 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH6197506 Supplier Thermo Scientific Chemicals Supplier No. H6197506
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 6319-40-0 | C7H4BrNO4 | 246.016 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 6319-40-0
Molecular Formula C7H4BrNO4
Molecular Weight (g/mol) 246.016
MDL Number MFCD00272137
InChI Key RVCTZJVBWNFYRU-UHFFFAOYSA-N
Synonym 4-bromo-3-nitrobenzoic acid, 3-nitro-4-bromobenzoic acid, benzoic acid, 4-bromo-3-nitro, 4-bromo-3-nitro-benzoic acid, zlchem 531, pubchem2194, acmc-209ndq, ksc353s1b, 4-bromo-3-nitrobenzoic acid #
PubChem CID 232986
SMILES C1=CC(=C(C=C1C(=O)O)[N+](=O)[O-])Br

Specifications

Melting Point 199°C to 206°C
Color White to Yellow
Quantity 5 g
Formula Weight 246.01
Percent Purity 95%
Physical Form Solid
Chemical Name or Material 4-Bromo-3-nitrobenzoic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only