missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylpyrazole 98.0+%, TCI America™

Catalog No. B35221G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B35221G 1 g
B35225G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B35221G Supplier TCI America Supplier No. B35221G

Chemical Identifiers

CAS 13808-64-5
Molecular Formula C4H5BrN2
Molecular Weight (g/mol) 161.002
MDL Number MFCD00005241
InChI Key IXQPRETWBGVNPJ-UHFFFAOYSA-N
Synonym 4-bromo-3-methylpyrazole, 4-bromo-3-methyl-1h-pyrazole, 3-methyl-4-bromopyrazole, 1h-pyrazole, 4-bromo-3-methyl, 4-bromo-3-methyl-2h-pyrazole, 4-bromo-3-methyl pyrazole, 3-methyl-4-bromo-1h-pyrazole, pubchem22533, timtec-bb sbb003957, acmc-1buv4
PubChem CID 83741
IUPAC Name 4-bromo-5-methyl-1H-pyrazole
SMILES CC1=C(C=NN1)Br

Specifications

Melting Point 76°C
Color White-Yellow
Quantity 1 g
Formula Weight 161.00
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material 4-Bromo-3-methylpyrazole