missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-3-methylanisole, 97%

Catalog No. p-7082065
Change view
Click to view available options
Quantity:
10 g
50 g
250 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1231809 10 g
AAA1231818 50 g
AAA1231830 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1231809 Supplier Thermo Scientific Chemicals Supplier No. A1231809

CAS: 27060-75-9 | C8H9BrO | 201.063 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 27060-75-9
Molecular Formula C8H9BrO
Molecular Weight (g/mol) 201.063
MDL Number MFCD00060687
InChI Key BLZNSXFQRKVFRP-UHFFFAOYSA-N
Synonym 2-bromo-5-methoxytoluene, 4-bromo-3-methylanisole, benzene, 1-bromo-4-methoxy-2-methyl, 4-bromo-3-methyl-anisole, unii-okr329132j, 1-bromo-2-methyl-4-methoxybenzene, 1-brom-4-methoxy-2-methylbenzol, pubchem3796, 3-methyl-4-bromoanisole, acmc-1cg4z
PubChem CID 117915
IUPAC Name 1-bromo-4-methoxy-2-methylbenzene
SMILES CC1=C(C=CC(=C1)OC)Br

Specifications

Density 1.45
Boiling Point 111°C to 112°C (15 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.561
Quantity 10 g
Beilstein 1933865
Formula Weight 201.06
Percent Purity 97%
Chemical Name or Material 4-Bromo-3-methylanisole
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only